Order Entry
Ireland
ContactUsLinkComponent
Potassium disulphite
Corrosive
Irritant
Potassium disulphite
Catalog # ROTHP019.1
Supplier:  Roth Carl
CAS Number:  
Corrosive
Irritant
Potassium disulphite
Catalog # ROTHP019.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :222.33 g/mol
Merck Index:13,07729
Melting Pt:190 °C
Density:1,2 g/cm³ (20 °C)
EINECS:240-795-3
 :GHS05
 :GHS07
CAS Number:16731-55-8
Formula:K₂O₅S₂
MDL Number:MFCD00167605