Disodium hydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7558-79-4
:
Formula:Na₂HPO₄
MDL Number:MFCD00003496
:141.96 g/mol
Merck Index:12,08805
Melting Pt:250 °C
Density:1,53 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
Frequently Bought Together