di-Potassium hydrogen phosphate
Catalog # MOLE33750632-5KG
Supplier: Molekula
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
About this item
:
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383