Potassium hexacyanoferrate(III) ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:13746-66-2
:
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392
:329.25 g/mol
Merck Index:13,07712
Density:1,85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-323-3
Specification Test Results
- Assay:Min. 99.0 %
- Hexacyanoferrate (II) (as Fe(CN)6):Max. 0.05 %
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Pb (Lead):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC26810.232 spec_VWRC26810.232 spec_VWRC26810.298
Product specification
Frequently Bought Together