Potassium hexacyanoferrate(III) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13746-66-2
:
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392
:329.25 g/mol
Merck Index:14,7630
Melting Pt: Dec.
Density:1,89
Storage Temperature:Ambient
EINECS:237-323-3
Related Information
Frequently Bought Together