Potassium hexacyanoferrate(III) 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13746-66-2
:
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392
:329.25 g/mol
Merck Index:13,07712
Density:1,85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-323-3
Specification Test Results
- Appearance:Orange to red Crystalline powder or crystals
- Titration Iodometric:≥97.5 %
Specification Test Results
Frequently Bought Together