Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Water
Water
Water
STD33000
:
IONICS
:
Water
STD33000
:
IONICS
:
Pk:
1
Seq:
0001
:
:
18.02 g/mol
:
13,10098
:
100 °C (1013 hPa)
:
0 °C
:
1 g/cm³ (4 °C)
:
Ambient
:
231-791-2
:
7732-18-5
:
H₂O
:
MFCD00011332