

Molybdic acid for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-91-4
:
Formula:H₂MoO₄
MDL Number:MFCD00036287
Merck Index:13,06262
Boiling Pt:∼300 °C (1013 hPa)
Melting Pt:300 °C
Density:3,1 g/cm³
:161.95 g/mol
Storage Temperature:Ambient
EINECS:231-970-5
:GHS07
:GHS08
Specification Test Results
- Appearance:White to light yellow Powder
- Additional info:contains ammonium molybdate
- Titration with NaOH:≥85 % (as MoO3)
- Chloride (Cl):≤100 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤2000 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤30 ppm
Specification Test Results
Frequently Bought Together