2-Furaldehyde for synthesis, Sigma-Aldrich®
: Merck
: 2-Furfurol, Furfural
:98-01-1
:
:C₅H₄O₂
:MFCD00003229
:162
:–39 °C
:1.15 g/cm³ (20 °C)
:96.09 g/mol
:60
:202-627-7
:1199
:6.1 (3) II
:13,04325
:M-Clarity™ quality level = MQ200
:GHS06
:GHS08
:Ambient
- Environmentally Preferable:
- Solubility:83 g/l (20 °C)
- Melting Point:-37 °C
- Molar Mass:96.08 g/mol
- Boiling Point:162 °C (1013 hPa)
- Vapor Pressure:1 hPa (20 °C)
- Flash Point:60 °C
- Explosion Limit:2.1 - 19.3 %(V)
- Density:1.16 g/cm3 (20 °C)
- Ignition Point:315 °C
- Saturation concentration:5.8 g/m3 (20 °C) Air
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together