Propylcyclohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1678-92-8
:
Formula:C₉H₁₈
MDL Number:MFCD00013770
:126.24 g/mol
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–94,9 °C
Density:0,7936 g/cm³ (20 °C)
EINECS:216-836-6
ADR:3,III
:M-Clarity™ quality level = MQ100
UN:3295
Frequently Bought Together