
2,3-Dimethylquinoxaline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2379-55-7
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00006728
:158.2 g/mol
Boiling Pt:125…130 °C (1,5 mmHg)
Melting Pt:102…107 °C
Density:1,226 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:219-162-0
:GHS07
Frequently Bought Together