

Benzo[e]pyrene analytical standard, Supelco®
Catalog # 442475.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:252.32 g/mol
Merck Index:12,01135
Boiling Pt:492 °C (1013 hPa)
Melting Pt:178…179 °C
EINECS:205-892-7
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:192-97-2
Formula:C₂₀H₁₂