

4-Phenylaniline, Supelco®
Catalog # 31598-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:169.23 g/mol
Merck Index:12,01276
Boiling Pt:302 °C (1013 hPa)
Melting Pt:55 °C
Density:1,16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-177-1
:GHS08
:GHS07
CAS Number:92-67-1
Formula:C₁₂H₁₁N
MDL Number:MFCD00007879