Order Entry
Ireland
ContactUsLinkComponent
 
3,4,5-Trichlorobiphenyl (PCB No. 38)
Catalog # PROOCIL-PCB-38-C
CAS Number:  
3,4,5-Trichlorobiphenyl (PCB No. 38)
Catalog # PROOCIL-PCB-38-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

 :
 :257.55 g/mol
CAS Number:53555-66-1
Formula:C₁₂H₇Cl₃