Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
122442A
:
AccuStandard
:
2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
122442A
:
AccuStandard
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
291.99 g/mol
:
Refrigerator
:
2315
:
9,II
:
35693-99-3
:
C₁₂H₆Cl₄