2,4,6-Triisopropylbenzenesulphonyl chloride
Catalog # MOLEM19580711
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:302.87 g/mol
Melting Pt:93…96 °C
Storage Temperature:Ambient
EINECS:229-479-6
UN:3261
ADR:8,II
:GHS05
CAS Number:6553-96-4
Formula:C₁₅H₂₃ClO₂S
MDL Number:MFCD00007433