Order Entry
Ireland
ContactUsLinkComponent
 
2,4,6-Trimethylthiophenol ≥97%
Catalog # B24798.03
CAS Number:  
2,4,6-Trimethylthiophenol ≥97%
Catalog # B24798.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.26 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1541-10-2
Formula:C₉H₁₂S
MDL Number:MFCD00218475