2,4,6-Trimethylbenzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:938-18-1
:
Formula:C₁₀H₁₁ClO
MDL Number:MFCD00013650
:182.65 g/mol
Boiling Pt:143…146 °C (60 mmHg)
Density:1,0967 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:213-339-6
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5280 to 1.5300 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together