
2,4-Dimethylbenzoyl chloride ≥97%
Catalog # H50387.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:21900-42-5
:
Formula:C₉H₉ClO
:168.62 g/mol
Boiling Pt:82 °C (1 mmHg)
Melting Pt:24…26 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
MDL Number:MFCD00798005