
2,6-Dimethylphenylboronic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100379-00-8
:
Formula:C₈H₁₁BO₂
MDL Number:MFCD01009693
:149.98 g/mol
Melting Pt:105 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together