Phenolphthalein, GPR RECTAPUR®
Catalog # 26237.231
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Suited for pH indicator:Passes test
- Melting point:259 - 264 °C
- Ignition residue (SO4):Max. 0.1 %
- Insolubility in ethanol 96 % (1% W/V):Max. 50 ppm
Specification Test Results
About this item
:
:318.33 g/mol
Merck Index:14,07243
Melting Pt:258…262 °C
Density:1,3 g/cm³ (20 °C)
EINECS:201-004-7
REACH:01-2119498295-24
:GHS08
Storage Temperature:Ambient
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913