Benzophenone 99%, pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:13,01099
- Boiling Pt:305 °C (760 mmHg)
- Melting Pt:47…49 °C
- Density:1,085 g/cm³ (20 °C)
- Flash Pt:143 °C
- Storage Temperature:Ambient
- EINECS:204-337-6
- ADR:9,III
3 Options Available Below
Product Details & Documents
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:13,01099
- Boiling Pt:305 °C (760 mmHg)
- Melting Pt:47…49 °C
- Density:1,085 g/cm³ (20 °C)
- Flash Pt:143 °C
- Storage Temperature:Ambient
- EINECS:204-337-6
- ADR:9,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:47 to 49 °C
- GC:≥98.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



