Bromophenol blue ACS, Reag. Ph. Eur., Supelco®
: Merck
:115-39-9
:
:C₁₉H₁₀Br₄O₅S
:MFCD00005875
:669.97 g/mol
:13,01429
:273 °C
:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:204-086-2
- Solubility:(20 °C) practically insoluble
- Melting Point:273 °C (decomposition)
- Molar Mass:669.96 g/mol
- Bulk Density:730 kg/m3
Frequently Bought Together