Order Entry
Ireland
ContactUsLinkComponent
Bromophenol blue Reag. Ph. Eur. indicator, Fluka™
Bromophenol blue Reag. Ph. Eur. indicator, Fluka™
Certificates
Bromophenol blue Reag. Ph. Eur. indicator, Fluka™
Certificates
Specification Test Results
  • Indicator solution:
    complying
  • Test of sensitivity:
    complying
  • Change range (pH 2,8-4,4):
    complying
  • UV spectrum:
    complying