Order Entry
Ireland
ContactUsLinkComponent
Sodium toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Irritant
Sodium toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Sodium toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    194.18 g/mol
  • Synonyms:
    4-Toluenesulfonic acid sodium salt
  • Hill Formula:
    C7H7NaO3S
  • Chemical Formula:
    4-(CH3)C6H4SO3Na

Specification Test Results

Related Information

Frequently Bought Together