

2-tert-Butylhydroquinone for synthesis, Sigma-Aldrich®
: Merck
:1948-33-0
:
:C₁₀H₁₄O₂
:MFCD00002344
:166.22 g/mol
:273 °C (1013 hPa)
:128 °C
:1,05 g/cm³ (20 °C)
:171 °C
:3077
:9,III
:M-Clarity™ quality level = MQ200
:217-752-2
:GHS07
:GHS09
:Ambient
- Melting Point:127 - 129 °C
- Molar Mass:166.21 g/mol
- Boiling Point:295 °C (1013 hPa)
- Flash Point:171 °C
- Density:1.05 g/cm3 (20 °C)
- Ignition Point:457 °C
Frequently Bought Together