o-Cresol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-48-7
:
Formula:C₇H₈O
MDL Number:MFCD00002226
:108.14 g/mol
Merck Index:13,02604
Boiling Pt:191 °C (760 mmHg)
Melting Pt:30…32 °C
Density:1,04 g/cm³
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:202-423-8
UN:3455
ADR:6.1,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Colorless to amber Low melting solid or liquid (May darken on storage)
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together