Order Entry
Ireland
ContactUsLinkComponent
 
Benzyl alcohol for synthesis, Sigma-Aldrich®
 :  Merck
 
undefined
Benzyl alcohol for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    40 g/l (20 °C)
  • Melting Point:
    -15.3 °C
  • Molar Mass:
    108.14 g/mol
  • Boiling Point:
    205 °C (1013 hPa)
  • Vapor Pressure:
    0.03 hPa (20 °C)
  • Flash Point:
    101 °C DIN 51758
  • Refractive Index:
    1.54
  • Explosion Limit:
    1.3 - 13 %(V)
  • Density:
    1.05 g/cm3 (20 °C)
  • Ignition Point:
    435 °C DIN 51794

 

Frequently Bought Together