Benzyl alcohol 99-100.5% (by GC), Puriss. NF, Ph. Eur., BP, meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Merck Index:13,01126
Boiling Pt:205,3 °C (1.013 hPa)
Melting Pt:–15,4 °C
Density:1,045 g/cm³ (20 °C)
Flash Pt:100,4 °C
EINECS:202-859-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Ignition Residue:≤0.005% (as SO₄)
- Identity (IR):complying
- Assay (GC):99.0 - 100.5 %
- Density (D 20/20):1.043 - 1.049
- Density (D 25/25):1.042 - 1.047
- Refractive index (n 20/D):1.539 - 1.541
- Non-volatile matter:Max. 0.05 %
- Water (Karl Fischer):Max. 0.1 %
- Sulfated ash:Max. 0.005 %
- Free acid:complying
- Free acid (as C₆H₅COOH):Max. 0.02 %
- Benzaldehyde (GC):Max. 0.05 %
- Cyclohexylmethanol (GC):Max. 0.10 %
- Sum peaks before Benzyl alcohol (GC):Max. 0.02 %
- Sum peaks after Benzyl alcohol (GC):Max. 0.2 %
- Related substances (GC):complying
- Peroxide value:Max. 5
- Appearance of the solution:complying
- Residual solvents:complying
- Vapor Density:3.7 (vs air)
Specification Test Results
Frequently Bought Together