2-tert-Butyl-4-methoxyphenol for synthesis, Sigma-Aldrich®
: Merck
: 3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole
:121-00-6
:
:C₁₁H₁₆O₂
:MFCD00040484
:267…269 °C (1013 hPa)
:60…61 °C
:1.0587 g/cm³ (20 °C)
:180.25 g/mol
:M-Clarity™ quality level = MQ200
:204-442-7
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) insoluble
- Melting Point:48 - 63 °C
- Molar Mass:180.25 g/mol
- Boiling Point:263 °C (1013 hPa)
- Vapor Pressure:1.33 hPa (105 °C)
- Flash Point:130 °C
Frequently Bought Together