


2,4-Diaminotoluene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-80-7
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00007804
:122.17 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:99 °C
Density:1,045 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-453-1
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:1709
:GHS06
:GHS08
:GHS09
Specification Test Results
- Solubility:35 g/l (20 °C)
- Melting Point:96 - 99 °C
- Molar Mass:122.17 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:284 - 286 °C (1013 hPa)
- Vapor Pressure:1 hPa (106 °C)
- Flash Point:169 °C
- Ignition Point:520 °C
Specification Test Results
Frequently Bought Together