α,α,α,2,4,6-Hexafluorotoluene 96%
Catalog # L18167.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:122030-04-0
:
Formula:C₇H₂F₆
MDL Number:MFCD00190109
:200.08 g/mol
Boiling Pt:105…107 °C
Density:1,475
Flash Pt:22 °C (71 °F)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS07