α-Bromo-4-chloro-3-fluorotoluene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:206362-80-3
:
Formula:C₇H₅BrClF
MDL Number:MFCD04115859
Melting Pt:30…33 °C
UN:3261
ADR:8,II
:223.47 g/mol
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together