

Benzenesulphonyl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-09-9
:
Formula:C₆H₅ClO₂S
MDL Number:MFCD00007426
:176.62 g/mol
Merck Index:13,01072
Boiling Pt:251 °C (1013 hPa)
Melting Pt:15 °C
Density:1,3842 g/cm³ (20 °C)
EINECS:202-636-6
UN:2225
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:119 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble,(decomposition)
- Melting Point:15 °C
- Molar Mass:176.62 g/mol
- Boiling Point:252 °C (1013 hPa) decomposes
- Vapor Pressure:6 hPa (20 °C)
- Flash Point:123 °C
- Refractive Index:1.553 (20 °C, 589 nm)
- Density:1.38 g/cm3 (20 °C)
- Ignition Point:450 °C
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Frequently Bought Together