
Diphenyl ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:101-84-8
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00003034
:170.21 g/mol
Merck Index:12,07442
Boiling Pt:498 °C (1013 hPa)
Melting Pt:24…27 °C
Density:1,07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-981-2
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:125 °C
:GHS09
Specification Test Results
- Solubility:0.018 g/l (25 °C)
- Melting Point:24 - 27 °C
- Molar Mass:170.21 g/mol
- Boiling Point:259 °C (1013 hPa)
- Vapor Pressure:0.05 hPa (20 °C)
- Flash Point:115 °C
- Refractive Index:1.5785 (25 °C, 589 nm)
- Explosion Limit:0.8 - 1.5 %(V)
- Density:1.07 g/cm3 (20 °C)
- Ignition Point:610 °C
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Frequently Bought Together