Hydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-31-9
:
Formula:C₆H₄(OH)₂
MDL Number:MFCD00002339
:110.11 g/mol
Merck Index:13,04833
Boiling Pt:285 °C (1013 hPa)
Melting Pt:172 °C
Density:1,332 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-617-8
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:165 °C
:GHS05
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:70 g/l (25 °C)
- Melting Point:172 °C (decomposition)
- Molar Mass:110.11 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:287 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (25 °C)
- Flash Point:165 °C
- Density:1.358 g/cm3 (20 °C)
- pH:(70 g/l, H2O, 20 °C)
- Ignition Point:515 °C
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22333.1000_TDS_EN ProductPage_822333
Product specification
msds_822333
SDS
Frequently Bought Together