Allyl phenyl ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1746-13-0
:
Formula:H₂CCHCH₂OC₆H₅
MDL Number:MFCD00008644
:134.18 g/mol
Boiling Pt:191…192 °C
Density:0,979
Flash Pt:62 °C (143 °F)
Storage Temperature:Ambient
EINECS:217-125-3
Related Information
Frequently Bought Together