Order Entry
Ireland
ContactUsLinkComponent
Diphenylamine ACS
Toxic
Health
Environment
Diphenylamine ACS
Certificates
undefined
Toxic
Health
Environment
Diphenylamine ACS

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    52.5 to 54.0 °C
  • Solubility:
    (in alcohol) Passes test
  • Residue after ignition:
    ≤0.03 %
  • Sensitivity:
    (to nitrate) Passes test
  • Nitrate (NO3):
    Passes test

Specification Test Results

Related Information

Frequently Bought Together