p-Anisidine 99%
: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline
:104-94-9
:
:C₇H₉NO
:MFCD00007864
:243
:56…59 °C
:1.06 g/cm³
:123.15 g/mol
:122
:Ambient
:2811
:6.1 III
:13,00670
:203-254-2
:GHS06
:GHS08
:GHS09
- Environmentally Preferable:
- Appearance:Grey to brown Crystalline powder or granules or flakes
- Infrared spectrum:Conforms
- Melting point:56 to 59 °C
- GC:≥98.5 %
- Stabilizer:10 ppm Kerobit(N,N'-Di-sec-butyl-p-phenylenediamine)
- Pyrrolidone:≤0.2 %
Frequently Bought Together