2-Iodothioanisole
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: (2-Iodophenyl)(methyl)sulphane, 2-Iodophenyl methyl sulphide, 1-Iodo-2-(methylthio)benzene
CAS Number:33775-94-9
:
Formula:C₇H₇IS
MDL Number:MFCD00051611
Boiling Pt:173 °C (20 mmHg)
Molecular Weight:250.1 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together