Potassium hydrogen phthalate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
CAS Number:877-24-7
:
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070
Density:1.6362 g/cm³ (20 °C)
Molecular Weight:204.22 g/mol
Melting Pt:295…300 °C
Merck Index:13,07694
Storage Temperature:Ambient
EINECS:212-889-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together