Order Entry
Ireland
ContactUsLinkComponent
p-Anisic acid
p-Anisic acid
Catalog # ROTH3631.1
Supplier:  Roth Carl
CAS Number:  
p-Anisic acid
Catalog # ROTH3631.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.15 g/mol
Merck Index:13,00669
Boiling Pt:275 °C (1013 hPa)
Melting Pt:182…185 °C
Density:∼1,385 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-818-5
CAS Number:100-09-4
Formula:C₈H₈O₃
MDL Number:MFCD00002542