Order Entry
Ireland
ContactUsLinkComponent
Methyl-4-hydroxybenzoate sodium salt
Irritant
Methyl-4-hydroxybenzoate sodium salt
Catalog # PROOMM0431.01
CAS Number:  
Irritant
Methyl-4-hydroxybenzoate sodium salt
Catalog # PROOMM0431.01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :174.13 g/mol
Melting Pt:> 125 °C
EINECS:225-714-1
 :GHS07
Storage Temperature:Ambient
CAS Number:5026-62-0
Formula:C₈H₇NaO₃
MDL Number:MFCD00016470