
Methyl-4-hydroxybenzoate sodium salt, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:5026-62-0
:
Formula:C₈H₇NaO₃
MDL Number:MFCD00016470
:174.13 g/mol
Melting Pt:> 125 °C
EINECS:225-714-1
:GHS05
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMERP1.06756.5000_TDS_EN ProductPage_106756
Product specification
msds_106756
SDS
Frequently Bought Together