
Methyl-4-hydroxybenzoate sodium salt 99%
ACRO235145000
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Appearance:White to almost white Powder or crystalline powder
- Infrared spectrum:Conforms
- Water:≤5 % (K.F.)
- Titration with HClO4:98.5 to 102.0 % (On anhydrous substance)
:
:174.13 g/mol
:>125 °C
:>100 °C
:225-714-1
:GHS05
:Ambient
:5026-62-0
:C₈H₇NaO₃
:MFCD00016470