Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propyl 4-hydroxybenzoate
Propyl-4-hydroxybenzoate
Propyl-4-hydroxybenzoate
Propyl-4-hydroxybenzoate
ROTH4749.1
:
Roth Carl
:
Propyl-4-hydroxybenzoate
ROTH4749.1
:
Roth Carl
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
180.2 g/mol
:
13,07958
:
292 °C (1013 hPa)
:
95 °C
:
1,28 g/cm³ (20 °C)
:
Ambient
:
202-307-7
:
GHS07
:
94-13-3
:
C₁₀H₁₂O₃
:
MFCD00002354