Order Entry
Ireland
ContactUsLinkComponent
 
 
 
 
Chlorobenzene 99%, Reagent Grade
  101389-2.5L
 :  
undefined
 
 
 
Chlorobenzene 99%, Reagent Grade
  101389-2.5L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Identity (IR):
    complying
  • Assay (GC):
    min. 98.5 %

 

 

 :
 :112.56 g/mol
 :13,02139
 :130-132 °C (1,013 hPa)
 :130…132 °C (1,013 hPa)
 :-45 °C
 :–45 °C
 :1,107…1.109 g/cm³ (20 °C)
 :28 °C
 :Ambient
 :203-628-5
 :1134
 :3,III
 :01-2119432722-45
 :GHS07
 :GHS02
 :GHS09
 :108-90-7
 :C₆H₅Cl
 :MFCD00000530