Order Entry
Ireland
ContactUsLinkComponent
p-Anisaldehyde
p-Anisaldehyde
Catalog # ROTH7201.1
Supplier:  Roth Carl
CAS Number:  
p-Anisaldehyde
Catalog # ROTH7201.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,00666
Boiling Pt:140 °C (1013 hPa)
Melting Pt:0…2 °C
Density:1,12 g/cm³ (25 °C)
Flash Pt:110 °C
 :136.15 g/mol
Storage Temperature:Ambient
EINECS:204-602-6
CAS Number:123-11-5
Formula:C₈H₈O₂
MDL Number:MFCD00003385