2-Bromo-5-fluoroaniline
Catalog # MOLE32061408
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:6.1,III
:190.01 g/mol
UN:2811
:GHS07
CAS Number:1003-99-2
Formula:C₆H₅BrFN
MDL Number:MFCD00070750