Order Entry
Ireland
ContactUsLinkComponent
1-Bromo-2-chloro-4-fluorobenzene ≥98%
1-Bromo-2-chloro-4-fluorobenzene ≥98%
Catalog # B25538.06
CAS Number:  
1-Bromo-2-chloro-4-fluorobenzene ≥98%
Catalog # B25538.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :209.44 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:110407-59-5
Formula:C₆H₃BrClF
MDL Number:MFCD00142587